Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000247.2 | NP_000238.1 | 3 | 642 | ||
GenBank mRNA | AB506764.1 | - | 3 | 576 | ||
AK094237.1 | - | 3 | 1314 | |||
AY204547.1 | - | 3 | 576 | |||
AY750850.1 | - | 3 | 576 | |||
BC016929.1 | - | 3 | 630 | 4446575 | ||
L14848.1 | - | 3 | 615 |