Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHKB-CPT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027928.2 | 26 | 3549 | |||
GenBank mRNA | AB051457.1 | - | 23 | 4187 | ||
BC131570.1 | - | 18 | 2329 | 40108355 |
Gene Symbol: CPT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145134.1 | NP_001138606.1 | 15 | 2020 | ||
NM_001145135.1 | NP_001138607.1 | 16 | 2122 | |||
NM_001145137.1 | NP_001138609.1 | 15 | 2146 | |||
NM_004377.3 | NP_004368.1 | 16 | 2083 | |||
NM_152245.2 | NP_689451.1 | 16 | 2122 | |||
NM_152246.2 | NP_689452.1 | 16 | 2083 | |||
GenBank mRNA | AK223174.1 | - | 16 | 2022 | ||
AK291316.1 | - | 16 | 2047 | |||
AK297866.1 | - | 15 | 1932 | |||
AK299395.1 | - | 16 | 1816 | |||
BC142954.1 | - | 15 | 2089 | 8860341 | ||
D87812.1 | - | 15 | 2003 | |||
U62733.1 | - | 15 | 2030 | |||
U66828.1 | - | 16 | 2022 | |||
Y08682.1 | - | 16 | 2035 | |||
Y08683.1 | - | 16 | 2035 |