Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC23L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317949.1 | NP_001304878.1 | 7 | 745 | ||
NM_144725.3 | NP_653326.3 | 7 | 810 | |||
XM_011513984.2 | XP_011512286.1 | 7 | 747 | |||
XM_011513985.2 | XP_011512287.1 | 7 | 747 | |||
XM_011513987.2 | XP_011512289.1 | 7 | 747 | |||
XM_017009119.1 | XP_016864608.1 | 7 | 991 | |||
XM_017009120.1 | XP_016864609.1 | 7 | 810 | |||
XM_017009121.1 | XP_016864610.1 | 7 | 855 | |||
XM_017009122.1 | XP_016864611.1 | 7 | 926 | |||
XM_017009123.1 | XP_016864612.1 | 7 | 747 | |||
XM_017009124.1 | XP_016864613.1 | 5 | 687 | |||
XM_017009125.1 | XP_016864614.1 | 5 | 488 | |||
XM_017009126.1 | XP_016864615.1 | 5 | 621 | |||
GenBank mRNA | AK058168.1 | - | 7 | 810 | ||
AK293829.1 | - | 5 | 506 | |||
AY498865.1 | - | 6 | 707 | |||
BC057775.1 | - | 7 | 782 | 5295348 | ||
BX088767.1 | - | 2 | 87 | |||
JN799997.1 | - | 7 | 745 |