Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207305.4 | NP_997188.2 | 1 | 222 | ||
GenBank mRNA | BC089432.1 | - | 1 | 224 | 30555309 | |
CF139066.1 | - | 1 | 241 |