Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015131.2 | NP_055946.1 | 18 | 2797 | ||
GenBank mRNA | BC024234.1 | - | 4 | 1113 | ||
D26488.1 | - | 18 | 2770 | |||
D87716.1 | - | 16 | 2278 |