Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098812.1 | NP_001092282.1 | 10 | 1839 | ||
NM_001098813.1 | NP_001092283.1 | 10 | 2033 | |||
NM_001300798.1 | NP_001287727.1 | 10 | 1833 | |||
NM_001300799.1 | NP_001287728.1 | 10 | 2101 | |||
NM_015146.1 | NP_055961.1 | 10 | 2107 | |||
XM_017009258.1 | XP_016864747.1 | 10 | 1908 | |||
XM_017009259.1 | XP_016864748.1 | 10 | 1902 | |||
XM_017009260.1 | XP_016864749.1 | 10 | 1855 | |||
XM_017009261.1 | XP_016864750.1 | 10 | 2170 | |||
GenBank mRNA | AF179995.1 | - | 10 | 2107 | ||
AF440761.1 | - | 10 | 2107 | |||
AF440763.1 | - | 10 | 1942 | |||
AK057797.1 | - | 10 | 1697 | |||
BC001329.1 | - | 7 | 1506 | |||
BC140759.1 | - | 10 | 1923 | 9021627 | ||
BC171779.1 | - | 10 | 1924 | 9054433 | ||
D86957.1 | - | 10 | 2107 |