Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4EBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004096.4 | NP_004087.1 | 1, 2 | 447 | ||
GenBank mRNA | BC005057.1 | - | 1, 2 | 173 | 2823322 | |
BC050633.1 | - | 1, 2 | 386 | 5555034 | ||
BG717474.1 | - | 1, 2 | 451 | |||
BT007317.1 | - | 1, 2 | 150 | |||
CR542240.1 | - | 1, 2 | 150 | |||
L36056.1 | - | 1, 2 | 150 |