Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015473.3 | NP_056288.2 | 10, 11 | 1785 | ||
GenBank mRNA | AB037737.1 | - | 1, 2 | 233 | ||
AK092400.1 | - | 3, 4 | 527 | |||
BC045764.1 | - | 4, 5 | 798 | |||
CR749444.1 | - | 3, 4 | 483 |