Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM169A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015566.2 | NP_056381.1 | 13 | 2625 | ||
NR_046462.1 | 12 | 2588 | ||||
XM_005248480.2 | XP_005248537.1 | 13 | 2760 | |||
XM_011543306.2 | XP_011541608.1 | 13 | 2847 | |||
XM_011543307.2 | XP_011541609.1 | 13 | 3276 | |||
XM_017009332.1 | XP_016864821.1 | 11 | 3523 | |||
XM_017009333.1 | XP_016864822.1 | 11 | 2497 | |||
GenBank mRNA | AB020695.1 | - | 13 | 2577 | ||
AK022981.1 | - | 4 | 1581 | |||
BX640902.1 | - | 12 | 2402 | |||
BX647536.1 | - | 12 | 2590 | |||
CR936791.1 | - | 13 | 2627 |