Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM169A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015566.2 | NP_056381.1 | 13 | 5149 | ||
NR_046462.1 | 12 | 5112 | ||||
XM_005248480.2 | XP_005248537.1 | 13 | 5284 | |||
XM_011543306.2 | XP_011541608.1 | 13 | 5371 | |||
XM_011543307.2 | XP_011541609.1 | 13 | 5800 | |||
XM_017009332.1 | XP_016864821.1 | 11 | 6047 | |||
XM_017009333.1 | XP_016864822.1 | 11 | 5021 | |||
GenBank mRNA | BC040298.1 | - | 1 | 947 | ||
BC044766.1 | - | 1 | 947 | |||
BX640902.1 | - | 12 | 4925 | |||
BX647536.1 | - | 12 | 5114 | |||
CR936791.1 | - | 13 | 5150 |