Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123366.1 | NP_001116838.1 | 3 | 353 | ||
XM_011525930.2 | XP_011524232.1 | 3 | 448 | |||
XM_017025710.1 | XP_016881199.1 | 3 | 448 | |||
GenBank mRNA | BM696260.1 | - | 3 | 353 |