Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002097.2 | NP_002088.2 | 8 | 1089 | ||
GenBank mRNA | AK057993.1 | - | 8 | 1625 | ||
BC143711.1 | - | 7 | 1081 | |||
D32257.1 | - | 8 | 1088 | |||
U14134.1 | - | 7 | 814 | |||
U20272.1 | - | 8 | 889 |