Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199380.1 | NP_001186309.1 | 11 | 2129 | ||
NM_001199381.1 | NP_001186310.1 | 11 | 2176 | |||
NM_001199382.1 | NP_001186311.1 | 11 | 2102 | |||
NM_001199383.1 | NP_001186312.1 | 11 | 2253 | |||
NM_144726.2 | NP_653327.1 | 11 | 2000 | |||
XM_017009138.1 | XP_016864627.1 | 11 | 2448 | |||
GenBank mRNA | AK056513.1 | - | 11 | 2000 | ||
AK075101.1 | - | 2 | 578 | |||
AK098802.1 | - | 6 | 1201 | |||
AK126290.1 | - | 1 | 914 | |||
AK292854.1 | - | 11 | 2242 | |||
AK294323.1 | - | 3 | 637 | |||
AK304228.1 | - | 11 | 2088 | |||
AK304435.1 | - | 11 | 2176 | |||
AK308394.1 | - | 11 | 2128 | |||
AL831829.1 | - | 1 | 264 | |||
BC042684.1 | - | 11 | 2265 | 4826810 | ||
BC044767.1 | - | 11 | 2265 | |||
BX537635.1 | - | 1 | 1560 |