Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282501.1 | NP_001269430.1 | 2 | 112 | ||
NM_001282502.1 | NP_001269431.1 | 2 | 112 | |||
NM_005361.3 | NP_005352.1 | 2 | 287 | |||
NM_175742.2 | NP_786884.1 | 4 | 273 | |||
NM_175743.2 | NP_786885.1 | 3 | 191 | |||
XM_017029519.1 | XP_016885008.1 | 3 | 249 | |||
GenBank mRNA | AK290443.1 | - | 2 | 112 | ||
BC108720.1 | - | 2 | 106 | 6065905 | ||
BG575302.1 | - | 3 | 136 | |||
BM781931.1 | - | 4 | 229 | |||
BX282627.1 | - | 3 | 151 |
Gene Symbol: MAGEA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321400.1 | NP_001308329.1 | 3 | 191 | ||
NM_001321401.1 | NP_001308330.1 | 4 | 273 | |||
NM_001321402.1 | NP_001308331.1 | 2 | 112 | |||
NM_001321403.1 | NP_001308332.1 | 2 | 112 | |||
XM_017029406.1 | XP_016884895.1 | 3 | 248 |