Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175742.2 | NP_786884.1 | 3 | 249 | ||
NM_175743.2 | NP_786885.1 | 2 | 167 | |||
XM_017029519.1 | XP_016885008.1 | 2 | 225 | |||
GenBank mRNA | BG575302.1 | - | 2 | 112 | ||
BM781931.1 | - | 3 | 205 | |||
BX282627.1 | - | 2 | 127 |
Gene Symbol: MAGEA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321400.1 | NP_001308329.1 | 2 | 167 | ||
NM_001321401.1 | NP_001308330.1 | 3 | 249 | |||
XM_017029406.1 | XP_016884895.1 | 2 | 224 |