Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019066.4 | NP_061939.3 | 1 | 3332 | ||
GenBank mRNA | AJ243531.1 | - | 1 | 1317 | ||
BC035839.1 | - | 1 | 1454 | |||
BC063834.1 | - | 1 | 1514 | |||
BC112257.1 | - | 1 | 1254 | |||
BC112259.1 | - | 1 | 1254 |