Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032962.4 | NP_116738.1 | 3 | 276 | ||
NM_032963.3 | NP_116739.1 | 2 | 228 | |||
GenBank mRNA | AK122606.1 | - | 1 | 1078 | ||
BC038289.1 | - | 2 | 193 | 5222914 | ||
BC045165.1 | - | 2 | 210 | 5225523 | ||
DA821276.1 | - | 2 | 230 | |||
Z49270.1 | - | 2 | 203 |
Gene Symbol: CCL15-CCL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027921.3 | 7 | 1186 | |||
NR_027922.3 | 6 | 1138 |