Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF185 SIRNA Assays ›
Gene Symbol:
ZNF185
Gene Name:
zinc finger protein 185 (LIM domain)
Gene Aliases:
SCELL
Chromosome Location:
Chr.X: 152898132 - 152973481 on Build GRCh38
UniGene ID:
Hs.16622
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s458566
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF185
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178106.1
NP_001171577.1
24
2887
NM_001178107.1
NP_001171578.1
23
2800
NM_001178108.1
NP_001171579.1
23
2794
NM_001178109.1
NP_001171580.1
22
2704
NM_001178110.1
NP_001171581.1
21
2614
NM_001178113.1
NP_001171584.1
15
2344
NM_001178115.1
NP_001171586.1
9
1695
NM_007150.3
NP_009081.2
23
2791
XM_005274730.2
XP_005274787.1
25
2994
XM_005274731.2
XP_005274788.1
25
2991
XM_005274735.2
XP_005274792.1
24
2904
XM_005274737.2
XP_005274794.1
24
2898
XM_005274738.2
XP_005274795.1
24
2895
XM_005274740.2
XP_005274797.1
23
2808
XM_005274741.2
XP_005274798.1
23
2808
XM_005274742.2
XP_005274799.1
23
2805
XM_005274744.2
XP_005274801.1
22
2721
XM_005274745.2
XP_005274802.1
22
2718
XM_005274746.3
XP_005274803.1
21
2637
XM_011531194.1
XP_011529496.1
23
2799
XM_011531195.1
XP_011529497.1
24
2796
XM_017029821.1
XP_016885310.1
26
3126
XM_017029822.1
XP_016885311.1
27
3146
XM_017029823.1
XP_016885312.1
26
3123
XM_017029824.1
XP_016885313.1
25
3042
XM_017029825.1
XP_016885314.1
25
3039
XM_017029826.1
XP_016885315.1
25
3036
XM_017029827.1
XP_016885316.1
25
3033
XM_017029828.1
XP_016885317.1
25
3030
XM_017029829.1
XP_016885318.1
24
2949
XM_017029830.1
XP_016885319.1
24
2943
XM_017029831.1
XP_016885320.1
24
2940
XM_017029832.1
XP_016885321.1
24
2931
XM_017029833.1
XP_016885322.1
23
2865
XM_017029834.1
XP_016885323.1
23
2853
XM_017029835.1
XP_016885324.1
23
2814
GenBank mRNA
AK056517.1
-
12
1850
AK095258.1
-
9
1667
AY997296.1
-
23
2780
BX537525.1
-
9
3369
BX640940.1
-
15
2091
Back To Top
More Information
Gene Ontology Categories:
Process(es)
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-2ddqc:80/100.66.79.182:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline