Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF185 SIRNA Assays ›
Gene Symbol:
ZNF185
Gene Name:
zinc finger protein 185 (LIM domain)
Gene Aliases:
SCELL
Chromosome Location:
Chr.X: 152898132 - 152973481 on Build GRCh38
UniGene ID:
Hs.16622
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s458567
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF185
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178106.1
NP_001171577.1
24
4220
NM_001178107.1
NP_001171578.1
23
4133
NM_001178108.1
NP_001171579.1
23
4127
NM_001178109.1
NP_001171580.1
22
4037
NM_001178110.1
NP_001171581.1
21
3947
NM_001178113.1
NP_001171584.1
15
3677
NM_001178115.1
NP_001171586.1
9
3028
NM_007150.3
NP_009081.2
23
4124
XM_005274730.2
XP_005274787.1
25
4327
XM_005274731.2
XP_005274788.1
25
4324
XM_005274735.2
XP_005274792.1
24
4237
XM_005274737.2
XP_005274794.1
24
4231
XM_005274738.2
XP_005274795.1
24
4228
XM_005274740.2
XP_005274797.1
23
4141
XM_005274741.2
XP_005274798.1
23
4141
XM_005274742.2
XP_005274799.1
23
4138
XM_005274744.2
XP_005274801.1
22
4054
XM_005274745.2
XP_005274802.1
22
4051
XM_005274746.3
XP_005274803.1
21
3970
XM_011531194.1
XP_011529496.1
23
4132
XM_011531195.1
XP_011529497.1
24
4129
XM_017029821.1
XP_016885310.1
26
4459
XM_017029822.1
XP_016885311.1
27
4479
XM_017029823.1
XP_016885312.1
26
4456
XM_017029824.1
XP_016885313.1
25
4375
XM_017029825.1
XP_016885314.1
25
4372
XM_017029826.1
XP_016885315.1
25
4369
XM_017029827.1
XP_016885316.1
25
4366
XM_017029828.1
XP_016885317.1
25
4363
XM_017029829.1
XP_016885318.1
24
4282
XM_017029830.1
XP_016885319.1
24
4276
XM_017029831.1
XP_016885320.1
24
4273
XM_017029832.1
XP_016885321.1
24
4264
XM_017029833.1
XP_016885322.1
23
4198
XM_017029834.1
XP_016885323.1
23
4186
XM_017029835.1
XP_016885324.1
23
4147
GenBank mRNA
AK056517.1
-
12
3183
AK095258.1
-
9
3000
AY997296.1
-
23
4113
BQ923456.1
-
1
433
BX537525.1
-
9
4703
BX640940.1
-
15
3424
Y09538.1
-
15
3412
Back To Top
More Information
Gene Ontology Categories:
Process(es)
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0