Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF185 SIRNA Assays ›
Gene Symbol:
ZNF185
Gene Name:
zinc finger protein 185 (LIM domain)
Gene Aliases:
SCELL
Chromosome Location:
Chr.X: 152898132 - 152973481 on Build GRCh38
UniGene ID:
Hs.16622
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s458568
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF185
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178106.1
NP_001171577.1
24
4341
NM_001178107.1
NP_001171578.1
23
4254
NM_001178108.1
NP_001171579.1
23
4248
NM_001178109.1
NP_001171580.1
22
4158
NM_001178110.1
NP_001171581.1
21
4068
NM_001178113.1
NP_001171584.1
15
3798
NM_001178115.1
NP_001171586.1
9
3149
NM_007150.3
NP_009081.2
23
4245
XM_005274730.2
XP_005274787.1
25
4448
XM_005274731.2
XP_005274788.1
25
4445
XM_005274735.2
XP_005274792.1
24
4358
XM_005274737.2
XP_005274794.1
24
4352
XM_005274738.2
XP_005274795.1
24
4349
XM_005274740.2
XP_005274797.1
23
4262
XM_005274741.2
XP_005274798.1
23
4262
XM_005274742.2
XP_005274799.1
23
4259
XM_005274744.2
XP_005274801.1
22
4175
XM_005274745.2
XP_005274802.1
22
4172
XM_005274746.3
XP_005274803.1
21
4091
XM_011531194.1
XP_011529496.1
23
4253
XM_011531195.1
XP_011529497.1
24
4250
XM_017029821.1
XP_016885310.1
26
4580
XM_017029822.1
XP_016885311.1
27
4600
XM_017029823.1
XP_016885312.1
26
4577
XM_017029824.1
XP_016885313.1
25
4496
XM_017029825.1
XP_016885314.1
25
4493
XM_017029826.1
XP_016885315.1
25
4490
XM_017029827.1
XP_016885316.1
25
4487
XM_017029828.1
XP_016885317.1
25
4484
XM_017029829.1
XP_016885318.1
24
4403
XM_017029830.1
XP_016885319.1
24
4397
XM_017029831.1
XP_016885320.1
24
4394
XM_017029832.1
XP_016885321.1
24
4385
XM_017029833.1
XP_016885322.1
23
4319
XM_017029834.1
XP_016885323.1
23
4307
XM_017029835.1
XP_016885324.1
23
4268
GenBank mRNA
AK056517.1
-
12
3304
AK095258.1
-
9
3121
AY997296.1
-
23
4234
BQ923456.1
-
1
554
BX537525.1
-
9
4824
BX640940.1
-
15
3545
Y09538.1
-
15
3533
Back To Top
More Information
Gene Ontology Categories:
Process(es)
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline