Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152553.4 | NP_689766.1 | 8 | 1109 | ||
XM_011535494.2 | XP_011533796.1 | 6 | 1750 | |||
XM_011535496.2 | XP_011533798.1 | 5 | 1524 | |||
XM_011535500.2 | XP_011533802.1 | 6 | 878 | |||
GenBank mRNA | BC026087.2 | - | 8 | 1105 | 4827114 |