Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277075.1 | NP_001264004.1 | 15 | 4352 | ||
XM_005259311.4 | XP_005259368.1 | 16 | 5292 | |||
XM_005259314.3 | XP_005259371.1 | 14 | 3378 | |||
XM_011527367.2 | XP_011525669.1 | 16 | 5307 | |||
XM_011527368.2 | XP_011525670.1 | 17 | 4645 | |||
XM_011527369.2 | XP_011525671.1 | 17 | 4649 | |||
XM_011527370.2 | XP_011525672.1 | 16 | 4506 | |||
XM_011527371.2 | XP_011525673.1 | 16 | 4493 | |||
XM_011527372.2 | XP_011525674.1 | 14 | 5142 | |||
XM_011527373.2 | XP_011525675.1 | 15 | 5133 | |||
XM_011527374.2 | XP_011525676.1 | 16 | 5142 | |||
XM_017027351.1 | XP_016882840.1 | 15 | 5166 | |||
GenBank mRNA | AL136846.1 | - | 13 | 2721 | ||
AL833862.1 | - | 7 | 2349 |