Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040214.2 | NP_001035304.1 | 2 | 501 | ||
NM_001300737.1 | NP_001287666.1 | 3 | 565 | |||
NM_001300738.1 | NP_001287667.1 | 2 | 501 | |||
NM_001300740.1 | NP_001287669.1 | 2 | 139 | |||
NM_153355.4 | NP_699186.2 | 2 | 501 | |||
XM_011535501.2 | XP_011533803.1 | 2 | 389 | |||
XM_011535502.2 | XP_011533804.1 | 2 | 156 | |||
XM_011535503.2 | XP_011533805.1 | 2 | 507 | |||
XM_017010318.1 | XP_016865807.1 | 2 | 505 | |||
XM_017010319.1 | XP_016865808.1 | 2 | 389 | |||
GenBank mRNA | AB070452.1 | - | 2 | 310 | ||
AK055326.1 | - | 2 | 134 | |||
AK308927.1 | - | 2 | 139 | |||
AK309802.1 | - | 2 | 139 | |||
AK310763.1 | - | 2 | 176 | |||
AK315886.1 | - | 2 | 140 | |||
BC035062.2 | - | 3 | 267 | 5264304 | ||
DB174077.1 | - | 1 | 176 |