Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040214.2 | NP_001035304.1 | 2 | 531 | ||
NM_001300737.1 | NP_001287666.1 | 3 | 595 | |||
NM_001300738.1 | NP_001287667.1 | 2 | 531 | |||
NM_001300740.1 | NP_001287669.1 | 2 | 169 | |||
NM_153355.4 | NP_699186.2 | 2 | 531 | |||
XM_011535501.2 | XP_011533803.1 | 2 | 419 | |||
XM_011535502.2 | XP_011533804.1 | 2 | 186 | |||
XM_011535503.2 | XP_011533805.1 | 2 | 537 | |||
XM_017010318.1 | XP_016865807.1 | 2 | 535 | |||
XM_017010319.1 | XP_016865808.1 | 2 | 419 | |||
GenBank mRNA | AB070452.1 | - | 2 | 340 | ||
AK055326.1 | - | 2 | 164 | |||
AK308927.1 | - | 2 | 169 | |||
AK309802.1 | - | 2 | 169 | |||
AK310763.1 | - | 2 | 206 | |||
AK315886.1 | - | 2 | 170 | |||
BC035062.2 | - | 3 | 297 | 5264304 | ||
DB174077.1 | - | 1 | 206 |