Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDXACB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138378.2 | NP_612387.1 | 5 | 2330 | ||
NR_038364.1 | 4 | 2173 | ||||
GenBank mRNA | AK300795.1 | - | 4 | 1804 | ||
BC006136.1 | - | 5 | 2009 |