Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197964.4 | NP_932068.2 | 2 | 429 | ||
NR_073058.1 | 3 | 302 | ||||
NR_073059.1 | 2 | 208 | ||||
NR_073060.1 | 2 | 188 | ||||
GenBank mRNA | AA393224.1 | - | 2 | 146 | ||
AF161386.1 | - | 2 | 235 | |||
BC008467.1 | - | 2 | 232 | 4276817 | ||
BI117784.1 | - | 2 | 429 | |||
BU852633.1 | - | 2 | 212 | |||
CB125801.1 | - | 2 | 410 |
Gene Symbol: C7orf55-LUC7L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244584.2 | NP_001231513.1 | 2 | 429 | ||
GenBank mRNA | AK297642.1 | - | 2 | 201 |