Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282855.1 | NP_001269784.1 | 11 | 2481 | ||
NM_001321293.1 | NP_001308222.1 | 11 | 2493 | |||
NM_178827.4 | NP_849149.3 | 11 | 2081 | |||
NR_104244.1 | 12 | 2264 | ||||
NR_104245.1 | 10 | 1905 | ||||
XM_005250161.3 | XP_005250218.1 | 12 | 2196 | |||
XM_005250162.4 | XP_005250219.1 | 11 | 2062 | |||
XM_011515833.2 | XP_011514135.1 | 12 | 2009 | |||
XM_011515834.2 | XP_011514136.1 | 11 | 2314 | |||
XM_017011771.1 | XP_016867260.1 | 12 | 1995 | |||
XM_017011772.1 | XP_016867261.1 | 6 | 1657 | |||
GenBank mRNA | AK093153.1 | - | 11 | 2001 | ||
BC026173.1 | - | 12 | 2268 | 4838069 | ||
BC032838.2 | - | 11 | 2006 | |||
BC036121.1 | - | 11 | 2481 | 5296826 | ||
BC068607.1 | - | 11 | 2006 | |||
BC091520.1 | - | 11 | 1936 | 30379054 | ||
BC128181.1 | - | 11 | 1980 | 40113310 | ||
BC128182.1 | - | 10 | 1804 | 40113312 |