Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007294.3 | NP_009225.1 | 12 | 4437 | ||
NM_007297.3 | NP_009228.2 | 11 | 4345 | |||
NM_007298.3 | NP_009229.2 | 11 | 915 | |||
NM_007299.3 | NP_009230.2 | 12 | 1090 | |||
NM_007300.3 | NP_009231.2 | 12 | 4437 | |||
NR_027676.1 | 12 | 4341 | ||||
GenBank mRNA | AF005068.1 | - | 12 | 4306 | ||
AK316200.1 | - | 11 | 987 | |||
AY751490.1 | - | 11 | 4205 | |||
BC046142.1 | - | 6 | 521 | |||
BC072418.1 | - | 12 | 1090 | |||
BC085615.1 | - | 12 | 995 | |||
BC115037.1 | - | 12 | 4342 | 7961446 | ||
JN686490.1 | - | 11 | 4205 | |||
U14680.1 | - | 12 | 4324 | |||
U64805.1 | - | 11 | 896 |