Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS37D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077621.1 | NP_001071089.1 | 4 | 1507 | ||
XM_017011779.1 | XP_016867268.1 | 4 | 1697 | |||
GenBank mRNA | AY081952.1 | - | 4 | 1507 | ||
BC064621.1 | - | 4 | 1354 |