Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194291.2 | NP_919267.2 | 2, 3 | 884 | ||
XM_011516847.2 | XP_011515149.1 | 2, 3 | 878 | |||
GenBank mRNA | BC032396.1 | - | 2, 3 | 185 | 5197382 | |
BC041379.1 | - | 2, 3 | 881 | 5275048 |