Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011540700.1 | XP_011539002.1 | 3, 4 | 291 | ||
GenBank mRNA | AK308458.1 | - | 1, 2 | 113 | ||
BC031250.1 | - | 3, 4 | 290 |