Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322019.1 | NP_001308948.1 | 8, 9 | 992 | ||
NM_004544.3 | NP_004535.1 | 8, 9 | 992 | |||
NR_136155.1 | 7, 8 | 4034 | ||||
NR_136157.1 | 6, 7 | 3865 | ||||
XM_011511228.2 | XP_011509530.1 | 8, 9 | 979 | |||
GenBank mRNA | AB209440.1 | - | 7, 8 | 4082 | ||
AF087661.1 | - | 8, 9 | 912 | |||
AF087854.1 | - | 8, 9 | 931 | |||
AF453834.1 | - | 9, 10 | 981 | |||
AK091719.1 | - | 3, 4 | 1993 | |||
AK290778.1 | - | 8, 9 | 914 | |||
BC003417.1 | - | 8, 9 | 926 | 3451514 |