Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRSAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005373.3 | NP_001005373.1 | 16 | 1479 | ||
NM_001005374.3 | NP_001005374.1 | 15 | 1184 | |||
NM_001190723.2 | NP_001177652.1 | 16 | 1479 | |||
NM_138361.5 | NP_612370.3 | 15 | 1738 | |||
XM_006717316.3 | XP_006717379.1 | 16 | 1479 | |||
XM_017015283.1 | XP_016870772.1 | 16 | 2478 | |||
XM_017015284.1 | XP_016870773.1 | 16 | 1474 | |||
GenBank mRNA | AK056203.1 | - | 16 | 1461 | ||
AK056305.1 | - | 4 | 324 | |||
AK091589.1 | - | 15 | 1486 | |||
AK226106.1 | - | 16 | 1465 | |||
AY358830.1 | - | 16 | 1444 | |||
BC009239.2 | - | 15 | 1184 | 2988664 |