Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM120AOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322224.1 | NP_001309153.1 | 3 | 925 | ||
NM_198841.2 | NP_942138.2 | 3 | 1854 | |||
NR_136229.1 | 3 | 1261 | ||||
NR_136230.1 | 4 | 1382 | ||||
NR_136231.1 | 4 | 1975 | ||||
NR_136232.1 | 4 | 1189 | ||||
NR_136233.1 | 3 | 1012 | ||||
NR_136234.1 | 4 | 1046 | ||||
NR_136235.1 | 3 | 1068 | ||||
NR_136236.1 | 3 | 1271 | ||||
NR_136237.1 | 4 | 1392 | ||||
NR_136238.1 | 4 | 1133 | ||||
GenBank mRNA | AK056096.1 | - | 3 | 1854 | ||
BC080634.1 | - | 4 | 916 | |||
BC107135.1 | - | 4 | 1047 | |||
BC107136.1 | - | 3 | 926 | |||
BC113074.1 | - | 3 | 926 |