Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM120AOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322224.1 | NP_001309153.1 | 2, 3 | 630 | ||
NM_198841.2 | NP_942138.2 | 2, 3 | 1559 | |||
NR_136229.1 | 2, 3 | 966 | ||||
NR_136230.1 | 2, 3 | 966 | ||||
NR_136231.1 | 2, 3 | 1559 | ||||
NR_136232.1 | 2, 3 | 773 | ||||
NR_136233.1 | 2, 3 | 717 | ||||
NR_136234.1 | 2, 3 | 630 | ||||
NR_136235.1 | 2, 3 | 773 | ||||
NR_136236.1 | 2, 3 | 976 | ||||
NR_136237.1 | 2, 3 | 976 | ||||
NR_136238.1 | 2, 3 | 717 | ||||
GenBank mRNA | AK056096.1 | - | 2, 3 | 1559 | ||
AK309269.1 | - | 2, 3 | 528 | |||
BC063650.1 | - | 2, 3 | 523 | |||
BC080634.1 | - | 2, 3 | 500 | |||
BC107135.1 | - | 2, 3 | 631 | |||
BC107136.1 | - | 2, 3 | 631 | |||
BC113074.1 | - | 2, 3 | 631 |