Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177704.1 | NP_001171175.1 | 7 | 1139 | ||
NM_144966.5 | NP_659403.4 | 31 | 6258 | |||
XM_005251382.3 | XP_005251439.1 | 30 | 6506 | |||
XM_005251384.4 | XP_005251441.1 | 9 | 1552 | |||
XM_006716729.3 | XP_006716792.1 | 10 | 1797 | |||
XM_011517758.2 | XP_011516060.1 | 7 | 1188 | |||
XM_017014316.1 | XP_016869805.1 | 30 | 6533 | |||
XM_017014317.1 | XP_016869806.1 | 31 | 6076 | |||
XM_017014318.1 | XP_016869807.1 | 31 | 6831 | |||
XM_017014319.1 | XP_016869808.1 | 32 | 6735 | |||
XM_017014320.1 | XP_016869809.1 | 31 | 6662 | |||
XM_017014321.1 | XP_016869810.1 | 30 | 6325 | |||
XM_017014322.1 | XP_016869811.1 | 30 | 6119 | |||
XM_017014323.1 | XP_016869812.1 | 30 | 5792 | |||
XM_017014325.1 | XP_016869814.1 | 30 | 6533 | |||
XM_017014326.1 | XP_016869815.1 | 29 | 5349 | |||
XM_017014327.1 | XP_016869816.1 | 26 | 4718 | |||
GenBank mRNA | AB160987.1 | - | 29 | 5442 | ||
BC031064.1 | - | 7 | 1139 | |||
BX537574.1 | - | 24 | 4833 | |||
BX641104.1 | - | 1 | 5763 |