Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA2026 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017969.2 | NP_001017969.2 | 8 | 4998 | ||
XM_005251385.2 | XP_005251442.1 | 7 | 5384 | |||
XM_006716730.3 | XP_006716793.1 | 7 | 3022 | |||
XM_011517759.2 | XP_011516061.1 | 8 | 5474 | |||
XM_011517760.2 | XP_011516062.1 | 8 | 5474 | |||
XM_011517761.2 | XP_011516063.1 | 5 | 2844 | |||
XM_017014331.1 | XP_016869820.1 | 8 | 5474 | |||
XM_017014332.1 | XP_016869821.1 | 4 | 2947 | |||
XM_017014333.1 | XP_016869822.1 | 5 | 2596 | |||
XM_017014334.1 | XP_016869823.1 | 4 | 2703 | |||
GenBank mRNA | AB095946.1 | - | 1 | 1047 | ||
AL833370.1 | - | 1 | 192 | |||
BC136846.1 | - | 3 | 2329 | 9020837 | ||
BX648372.1 | - | 4 | 2481 |