Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00961 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024283.1 | 2 | 1345 | |||
GenBank mRNA | AK056723.1 | - | 2 | 1346 | ||
BC034765.1 | - | 1 | 1344 |