Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080551.2 | NP_001074020.2 | 19 | 2969 | ||
NM_173521.4 | NP_775792.4 | 21 | 2976 | |||
NR_109816.1 | 22 | 3066 | ||||
XM_006716974.2 | XP_006717037.1 | 23 | 3162 | |||
XM_006716975.2 | XP_006717038.1 | 22 | 3057 | |||
XM_011518302.2 | XP_011516604.1 | 21 | 4971 | |||
XM_011518303.1 | XP_011516605.1 | 21 | 2943 | |||
XM_011518306.1 | XP_011516608.1 | 18 | 2563 | |||
XM_011518309.1 | XP_011516611.1 | 23 | 3162 | |||
XM_017014340.1 | XP_016869829.1 | 21 | 3780 | |||
XM_017014341.1 | XP_016869830.1 | 23 | 3162 | |||
GenBank mRNA | AK126055.1 | - | 19 | 2967 | ||
AL833535.1 | - | 21 | 2970 | |||
BC144680.1 | - | 22 | 2990 |