Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZXDB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007157.3 | NP_009088.1 | 1 | 3380 | ||
GenBank mRNA | AK124513.1 | - | 2 | 1378 |