Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AWAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002254.1 | NP_001002254.1 | 5 | 516 | ||
XM_011530876.2 | XP_011529178.1 | 5 | 548 | |||
GenBank mRNA | AY605053.1 | - | 5 | 516 |