Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXorf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144970.2 | NP_659407.1 | 5 | 735 | ||
XM_005272589.2 | XP_005272646.2 | 5 | 735 | |||
XM_006724528.2 | XP_006724591.1 | 4 | 580 | |||
XM_017029302.1 | XP_016884791.1 | 4 | 726 | |||
XM_017029303.1 | XP_016884792.1 | 4 | 580 | |||
XM_017029304.1 | XP_016884793.1 | 5 | 1897 | |||
XM_017029305.1 | XP_016884794.1 | 5 | 770 | |||
GenBank mRNA | AK095710.1 | - | 4 | 929 | ||
AK123989.1 | - | 5 | 583 | |||
AK304539.1 | - | 3 | 444 | |||
AK315384.1 | - | 5 | 713 | |||
AL832829.1 | - | 5 | 1879 | |||
BC025334.1 | - | 5 | 693 | 4559938 |