Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM122C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170779.1 | NP_001164250.1 | 4, 5 | 705 | ||
NM_138819.3 | NP_620174.1 | 4, 5 | 705 | |||
XM_005262382.1 | XP_005262439.1 | 5, 6 | 483 | |||
XM_005262383.2 | XP_005262440.1 | 5, 6 | 483 | |||
XM_005262384.4 | XP_005262441.1 | 5, 6 | 483 | |||
XM_005262386.3 | XP_005262443.1 | 5, 6 | 483 | |||
XM_006724734.3 | XP_006724797.1 | 5, 6 | 571 | |||
XM_006724735.1 | XP_006724798.1 | 5, 6 | 483 | |||
XM_006724736.2 | XP_006724799.1 | 5, 6 | 483 | |||
XM_011531295.1 | XP_011529597.1 | 5, 6 | 483 | |||
XM_011531296.2 | XP_011529598.1 | 5, 6 | 483 | |||
XM_011531297.2 | XP_011529599.1 | 5, 6 | 483 | |||
XM_011531298.2 | XP_011529600.1 | 5, 6 | 483 | |||
XM_011531299.1 | XP_011529601.1 | 2, 3 | 83 | |||
XM_011531300.2 | XP_011529602.1 | 2, 3 | 83 | |||
XM_011531301.2 | XP_011529603.1 | 2, 3 | 78 | |||
XM_011531302.2 | XP_011529604.1 | 2, 3 | 79 | |||
XM_017029308.1 | XP_016884797.1 | 5, 6 | 483 | |||
XM_017029309.1 | XP_016884798.1 | 2, 3 | 160 | |||
GenBank mRNA | AK130840.1 | - | 1, 2 | 41 | ||
AK295485.1 | - | 6, 7 | 1340 | |||
BC017868.1 | - | 4, 5 | 704 | 4699951 | ||
BC063491.1 | - | 4, 5 | 645 | 5229041 | ||
BG532414.1 | - | 704 |