Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMY1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303410.1 | NP_001290339.1 | 12 | 1519 | ||
NM_152585.2 | NP_689798.1 | 12 | 1524 | |||
XM_017030033.1 | XP_016885522.1 | 10 | 1229 | |||
GenBank mRNA | BC030018.2 | - | 12 | 1524 | 5269012 | |
X76060.1 | - | 12 | 1501 |
Gene Symbol: RBMY1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006121.3 | NP_001006121.1 | 12 | 1564 | ||
NM_001320948.1 | NP_001307877.1 | 11 | 1453 | |||
XM_011531433.1 | XP_011529735.1 | 9 | 1191 | |||
XM_011531434.1 | XP_011529736.1 | 11 | 1363 | |||
XM_017030057.1 | XP_016885546.1 | 10 | 1229 | |||
GenBank mRNA | BC140758.1 | - | 11 | 1366 | 9021626 |
Gene Symbol: RBMY1J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006117.3 | NP_001006117.2 | 12 | 1589 | ||
XM_011531500.1 | XP_011529802.1 | 11 | 1349 | |||
XM_011531502.1 | XP_011529804.1 | 9 | 1127 | |||
XM_011531503.1 | XP_011529805.1 | 9 | 1113 | |||
XM_017030059.1 | XP_016885548.1 | 10 | 1229 | |||
GenBank mRNA | AK313597.1 | - | 12 | 1593 | ||
BC152392.1 | - | 11 | 1434 | 8860211 |