Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145353.1 | NP_001138825.1 | 2 | 334 | ||
NM_172373.3 | NP_758961.1 | 3 | 479 | |||
XM_005266276.1 | XP_005266333.1 | 3 | 368 | |||
XM_005266277.3 | XP_005266334.1 | 3 | 911 | |||
XM_011534950.1 | XP_011533252.1 | 5 | 717 | |||
XM_011534951.2 | XP_011533253.1 | 4 | 1030 | |||
XM_017020409.1 | XP_016875898.1 | 5 | 697 | |||
XM_017020410.1 | XP_016875899.1 | 4 | 634 | |||
XM_017020411.1 | XP_016875900.1 | 4 | 580 | |||
XM_017020412.1 | XP_016875901.1 | 4 | 708 | |||
XM_017020413.1 | XP_016875902.1 | 4 | 762 | |||
GenBank mRNA | AK304289.1 | - | 2 | 277 | ||
BC030507.2 | - | 2 | 334 | 4385989 | ||
BQ722965.1 | - | 3 | 376 | |||
DC418141.1 | - | 3 | 480 | |||
M82882.1 | - | 3 | 421 |