Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMTC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181783.3 | NP_861448.2 | 14 | 2189 | ||
XM_011537980.2 | XP_011536282.1 | 13 | 1964 | |||
XM_011537981.2 | XP_011536283.1 | 7 | 853 | |||
GenBank mRNA | AJ697717.1 | - | 14 | 2059 | ||
AK074973.1 | - | 13 | 1816 | |||
AK290436.1 | - | 14 | 2189 | |||
AK302755.1 | - | 14 | 2189 | |||
BC117175.1 | - | 13 | 2010 | 40125726 | ||
BC117177.1 | - | 13 | 2011 | 40125728 | ||
BX538169.1 | - | 14 | 2162 | |||
CR749244.1 | - | 14 | 2141 | |||
CR749309.1 | - | 14 | 2176 | |||
CR933632.1 | - | 14 | 2140 | |||
CR936640.1 | - | 4 | 517 |