Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152589.1 | NP_689802.1 | 11 | 1140 | ||
XM_011537985.1 | XP_011536287.1 | 10 | 1266 | |||
XM_017018886.1 | XP_016874375.1 | 10 | 1077 | |||
XM_017018887.1 | XP_016874376.1 | 11 | 1383 | |||
XM_017018888.1 | XP_016874377.1 | 9 | 963 | |||
GenBank mRNA | AK093140.1 | - | 11 | 1140 | ||
AL833338.1 | - | 10 | 1266 |