Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMNTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145727.2 | NP_001139199.1 | 5 | 650 | ||
NM_001145728.2 | NP_001139200.1 | 7 | 1136 | |||
NM_001145729.1 | NP_001139201.1 | 6 | 1016 | |||
NM_001256266.1 | NP_001243195.1 | 7 | 1048 | |||
NM_152590.3 | NP_689803.2 | 6 | 1017 | |||
XM_011520575.1 | XP_011518877.1 | 9 | 1242 | |||
XM_011520576.2 | XP_011518878.1 | 9 | 1242 | |||
XM_011520577.2 | XP_011518879.1 | 9 | 1652 | |||
XM_011520578.2 | XP_011518880.1 | 9 | 1265 | |||
XM_011520579.2 | XP_011518881.1 | 9 | 1242 | |||
XM_011520580.1 | XP_011518882.1 | 8 | 1122 | |||
XM_011520582.1 | XP_011518884.1 | 9 | 1254 | |||
XM_011520583.1 | XP_011518885.1 | 8 | 1123 | |||
XM_011520584.1 | XP_011518886.1 | 6 | 930 | |||
XM_011520585.2 | XP_011518887.1 | 6 | 853 | |||
XM_011520587.1 | XP_011518889.1 | 6 | 626 | |||
XM_011520588.1 | XP_011518890.1 | 5 | 548 | |||
XM_017018891.1 | XP_016874380.1 | 8 | 1074 | |||
XM_017018892.1 | XP_016874381.1 | 8 | 1144 | |||
XM_017018893.1 | XP_016874382.1 | 7 | 953 | |||
GenBank mRNA | AK093323.1 | - | 6 | 978 | ||
AK296816.1 | - | 5 | 642 | |||
AK302292.1 | - | 7 | 1136 | |||
AK310411.1 | - | 6 | 1014 | |||
BC037957.1 | - | 7 | 1017 | 5744078 | ||
BP359528.1 | - | 4 | 469 | |||
DW009862.1 | - | 2 | 151 |