Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308339.1 | NP_001295268.1 | 5 | 783 | ||
NM_144973.3 | NP_659410.3 | 3 | 713 | |||
XM_005253326.2 | XP_005253383.1 | 4 | 715 | |||
XM_005253327.3 | XP_005253384.1 | 3 | 708 | |||
XM_006719040.3 | XP_006719103.1 | 4 | 715 | |||
XM_017018896.1 | XP_016874385.1 | 4 | 715 | |||
XM_017018897.1 | XP_016874386.1 | 4 | 1078 | |||
XM_017018898.1 | XP_016874387.1 | 4 | 715 | |||
GenBank mRNA | AK123642.1 | - | 1 | 130 | ||
AK125323.1 | - | 5 | 783 | |||
BC020855.1 | - | 2 | 383 | |||
BC068580.1 | - | 4 | 770 | 5265807 |