Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308339.1 | NP_001295268.1 | 11 | 2454 | ||
NM_144973.3 | NP_659410.3 | 9 | 2384 | |||
XM_005253326.2 | XP_005253383.1 | 11 | 2497 | |||
XM_005253327.3 | XP_005253384.1 | 10 | 2490 | |||
XM_005253330.3 | XP_005253387.1 | 11 | 1966 | |||
XM_006719040.3 | XP_006719103.1 | 10 | 2386 | |||
XM_011520590.2 | XP_011518892.1 | 8 | 1733 | |||
XM_017018896.1 | XP_016874385.1 | 11 | 2497 | |||
XM_017018897.1 | XP_016874386.1 | 11 | 2860 | |||
XM_017018898.1 | XP_016874387.1 | 10 | 1960 | |||
XM_017018899.1 | XP_016874388.1 | 10 | 1745 | |||
GenBank mRNA | AB209341.1 | - | 5 | 1014 | ||
AK095598.1 | - | 6 | 1195 | |||
AK125323.1 | - | 11 | 2454 | |||
BX537924.1 | - | 2 | 157 |